Learn More
Cole-Parmer DWK Life Sciences (Kimble) Kimax Serialized and Certified Class A Glass Burette, 25 mL; 1/Pk
Supplier: Cole-Parmer 3454102
Designed from ASTM E287-Class A requirements, this self-zeroing burette (buret) is permanantly marked with an individual serial number. Filling is completed thru the (insert image P/S) stopcock and accepts 1/4" ID flexible tubing for filling tube and overflow tube. Graduations are in black enamel.