Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSC Go Science Crazy Burette and Pipette Brush
Brush and clean small containers like burettes or pipettes. GSC International Burette and Pipette Brushes employ soft, natural bristles to avoid scratching and galvanized handles to prevent rust.
Supplier: GSC Go Science Crazy 410370DZ
Alert:
Intended for ages 13 and up.
Description
- Great for cleaning out burettes or pipettes in laboratory or classroom settings
- Small, natural bristles are soft and designed to clean small spaces without scratching
- Galvanized handles prevent rusting over repeated use
- Available in two sizes
Specifications
Soft | |
32 mm | |
760 mm |
Galvanized Steel | |
Cleaning burettes and pipettes | |
133 mm |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction