Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Burette Solution, Gas Analysis Chemcials, Burrell Scientific

Designed as the confining liquid in gas analysis

Manufacturer:  Burrell Scientific Inc 0398000016

Catalog No. 14-400-154


Add to cart

Description

Description

  • Designed as the confining liquid in gas analysis
  • For use in gas burettes, gas reservoirs, slow combustion, and explosion pipettes
  • Saturated acidified and colored

Recommended Storage

Store above 60°F

Warranty and Services

6 Month Limited Warranty

Specifications

Specifications

Burette Solution, Gas Analysis Chemcials
7664-93-9, 497-19-8
Sodium Acid Sulfate
Liquid
Less than water
Red
55%, 24%, 20.99%
>100°C (>212°F)
Water: Completely Soluble
1.18
340°C (644°F)
16 oz.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit