Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Burkholderia cenocepacia derived from ATCC™ BAA-245™

Simple, all-inclusive design making life easier for lab techs while reducing chances for errors. Each KWIK-STIK contains a qualitative lyophilized microorganism pellet, ampoule of hydrating fluid and inoculating swab.

$60.00 - $164.10

Specifications

Biosafety Level 2
ATCC® ATCC™ BAA-245
Format KWIK-STIK™
Packaging Plastic Canister
Shelf Life 8 Months (Minimum)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
23-047-353
View Documents
Microbiologics™
01269P
2 Pack Pack of 2 for $60.00
Add to cart
 
23-047-354
View Documents
Microbiologics™
01269K
6 Pack Pack of 6 for $164.10
Add to cart
 
Description

Description

  • Self-contained units of a single organism
Specifications

Specifications

2
KWIK-STIK™
8 Months (Minimum)
ATCC™ BAA-245
Plastic Canister
2 to 8°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit