Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Burrell Scientific Wrist Action™ Laboratory Shaker Accessory, Multi SUREGRIP™ Clamps
For use with Wrist Action™ Laboratory Shakers
$1160.00 - $1629.00
Catalog Number | Mfr. No. | No. of Positions | Weight (Metric) | Weight (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | No. of Positions | Weight (Metric) | Weight (English) | Price | Quantity & Availability | |||
14-400-141
|
Burrell Scientific
0757830400 |
4 | 907g | 2 lb. |
Each for $1,160.00
|
|
|||
14-400-142
|
Burrell Scientific
0757830800 |
8 | 1.8 kg | 4 lb. |
Each for $1,629.00
|
|
|||
Description
- Maximum adjustability and security
- Accommodates up to 4 vessels
- Holds up to 3 in./ 90mm containers
- Sold in pairs, shafts included
Specifications
Clamp | |
Wrist Action Shaker |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title