Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Burrell Scientific Wrist Action™ Laboratory Shaker Accessory, Snap and Lock Clamp

For use with Wrist Action Laboratory Shaker

Manufacturer:  Burrell Scientific 0757772000

Catalog No. 14-400-145


Add to cart

Description

Description

  • Maximum adjustability and security
  • Material: Aluminum
  • Color: Silver, red and black
Specifications

Specifications

Accommodates 1 Vessel
Wrist Action Laboratory Shaker
One year
Clamp
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit