Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Burrell Scientific Wrist Action™ Shaker-Magnum™ Laboratory Shakers

Designed to maintain constant swirling action as long as required for almost any laboratory application

Supplier:  Burrell Scientific 0757900445

 View more versions of this product

Catalog No. 14-400-138


Add to Cart

Includes: Side arm shafts, Clamps, Power cord, Operations manual, Allen Wrench, Lever arm brake

Description

Description

  • Industrial bench top shakers
  • Digital variable speed with timer
  • Material: Aluminum and steel
  • Max tested Load at 13.6kg (30 lb.)
  • Color: Silver and black

Warranty and Services

One Year

Specifications

Specifications

0 to 600 Oscillations/min.
Laboratory Shaker
147 lb.
100-AA
100/240 V
50/60 Hz
66.6 kg
4
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit