Learn More
PYREX™ Bushing Type Reducing Glass Joint Adapters
With standard taper inner and outer joints for changing standard taper joint sizes
Manufacturer:  PYREX™ 88252924
 View more versions of this product
Catalog No. 01-033G
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
These PYREX™ bushing adapters are fabricated with heavy walls for mechanical strength and are further strengthened by the heavy, uniform rim at the top which affords a serviceable finger grip. These adapters have a 34/45 Standard Taper outer and 24/40 Standard Taper inner joints that are shorter than the conventional type, thus allowing more compact and convenient assemblies. Useful for attaching thermometers with Standard Taper inner joints.
Specifications
Specifications
For attaching thermometers with standard taper inner joints
56 mm
Tube Adapter
2.19 in.
Pyrex™
29/42 Inner Bottom, 24/40 Outer Top