Learn More
PYREX™ Bushing Type Reducing Glass Joint Adapters
With standard taper inner and outer joints for changing standard taper joint sizes
Catalog Number | Mfr. No. | Standard Taper Joint No. | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Standard Taper Joint No. | Price | Quantity & Availability | |||||
01-033G
|
PYREX™
88252924 |
29/42 Inner Bottom, 24/40 Outer Top | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
These PYREX™ bushing adapters are fabricated with heavy walls for mechanical strength and are further strengthened by the heavy, uniform rim at the top which affords a serviceable finger grip. These adapters have a 34/45 Standard Taper outer and 24/40 Standard Taper inner joints that are shorter than the conventional type, thus allowing more compact and convenient assemblies. Useful for attaching thermometers with Standard Taper inner joints.