Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buspirone Hydrochloride, USP, 97.5-102.5%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
Packaging:
Amber Glass Bottle
33386-08-2
C21H32ClN5O2
421.97
RICLFGYGYQXUFH-UHFFFAOYSA-N
hydrogen 8-{4-[4-(pyrimidin-2-yl)piperazin-1-yl]butyl}-8-azaspiro[4.5]decane-7,9-dione chloride
[H+].[Cl-].O=C1CC2(CCCC2)CC(=O)N1CCCCN1CCN(CC1)C1=NC=CC=N1
33386-08-2
C21H32ClN5O2
421.97
RICLFGYGYQXUFH-UHFFFAOYSA-N
hydrogen 8-{4-[4-(pyrimidin-2-yl)piperazin-1-yl]butyl}-8-azaspiro[4.5]decane-7,9-dione chloride
[H+].[Cl-].O=C1CC2(CCCC2)CC(=O)N1CCCCN1CCN(CC1)C1=NC=CC=N1

B2850, 33386-08-2, C21H31N5O2•HCl

Spectrum™ Buspirone Hydrochloride, USP is an anti-anxiety agent and a serotonin receptor agonist belonging to the azaspirodecanedione class of compounds. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Quantity 5 g
Ignition Residue 0.005
CAS Max % 1
UN Number UN2811
Percent Purity 97.5 to 102.5%