Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butoxycarbonylmethyl butyl phthalate, 95%, ACROS Organics™

Click to view available options
Quantity:
100mL
Packaging:
Glass Bottle
85-70-1,84-74-2
C18H24O6
336.384
MFCD00053793
GOJCZVPJCKEBQV-UHFFFAOYSA-N
butyl phthalyl butyl glycolate,butoxycarbonylmethyl butyl phthalate,reomol 4pg,butyl carbobutoxymethyl phthalate,santicizer b-16,2-butoxy-2-oxoethyl butyl phthalate,butyl glycolyl butyl phthalate,butyl phthalate butyl glycolate,caswell no. 131b,dibutyl o-carboxybenzoyloxyacetate
6819
2-O-(2-butoxy-2-oxoethyl) 1-O-butyl benzene-1,2-dicarboxylate
CCCCOC(=O)COC(=O)C1=CC=CC=C1C(=O)OCCCC
85-70-1,84-74-2
C18H24O6
336.384
MFCD00053793
GOJCZVPJCKEBQV-UHFFFAOYSA-N
butyl phthalyl butyl glycolate,butoxycarbonylmethyl butyl phthalate,reomol 4pg,butyl carbobutoxymethyl phthalate,santicizer b-16,2-butoxy-2-oxoethyl butyl phthalate,butyl glycolyl butyl phthalate,butyl phthalate butyl glycolate,caswell no. 131b,dibutyl o-carboxybenzoyloxyacetate
6819
2-O-(2-butoxy-2-oxoethyl) 1-O-butyl benzene-1,2-dicarboxylate
CCCCOC(=O)COC(=O)C1=CC=CC=C1C(=O)OCCCC
Packaging Glass Bottle
Density 1.1000g/mL
Color Colorless
Quantity 100mL
Refractive Index 1.4900 to 1.4920 (20°C, 589nm)
Infrared Spectrum Authentic
Assay Percent Range 95%
Formula Weight 336.38
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material Butoxycarbonylmethyl butyl phthalate
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
May damage the unborn child. Suspected of damaging fertility.

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger