Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl Acetate, Exceeds A.C.S. Specifications, HPLC Grade, 99%, Spectrum™

Click to view available options
Quantity:
1 L
4 L
200 L
Packaging:
Amber Glass Bottle
Drum
123-86-4
123-86-4
Packaging Drum
Quantity 200 L
Water 0.0005
Titratable Acid 1.6 μeq/g
Residue after Evaporation 5 ppm
CAS Max % 1
Percent Purity 0.99
Grade HPLC

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.