Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl Alcohol, NF, 99.5%, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
71-36-3
C4H10O
74.12
LRHPLDYGYMQRHN-UHFFFAOYSA-N
butan-1-ol
CCCCO
71-36-3
C4H10O
74.12
LRHPLDYGYMQRHN-UHFFFAOYSA-N
butan-1-ol
CCCCO

B1197, 71-36-3, C4H10O

Spectrum™ Butyl Alcohol, NF is primarily a solvent and vehicle in many pharmaceutical formulations. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
UN Number UN1120
Percent Purity 99.5%
Non_volatile Matter 0.004%
Packaging Amber Glass Bottle
Quantity 4 L