Learn More
Butyl Alcohol, NF, 99.5%, Spectrum™
Click to view available options
Packaging:
Amber Glass Bottle
Drum
Steel Can
B1197, 71-36-3, C4H10O
Spectrum™ Butyl Alcohol, NF is primarily a solvent and vehicle in many pharmaceutical formulations. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Non_volatile Matter 4e-005
Packaging Steel Can