Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl Alcohol, PestiSolv(R), 99%, Spectrum™

Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass Bottle
71-36-3
71-36-3
CAS Max % 1
Percent Purity 0.99
Grade High Purity Solvent
Packaging Amber Glass Bottle
Water 0.0005
Quantity 1 L

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.