Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl chloroformate, 98%, ACROS Organics™

Click to view available options
Quantity:
25mL
1L
Packaging:
Glass bottle
592-34-7
C5H9ClO2
136.58
MFCD00000650
NRDQFWXVTPZZAZ-UHFFFAOYSA-N
butyl chloroformate,n-butyl chloroformate,butylchloroformate,carbonochloridic acid, butyl ester,chloroformic acid butyl ester,butyl chlorocarbonate,n-butyl chlorocarbonate,formic acid, chloro-, butyl ester,butyl-chloroformate,butylchloro-formate
61140
butyl carbonochloridate
CCCCOC(=O)Cl
592-34-7
C5H9ClO2
136.58
MFCD00000650
NRDQFWXVTPZZAZ-UHFFFAOYSA-N
butyl chloroformate,n-butyl chloroformate,butylchloroformate,carbonochloridic acid, butyl ester,chloroformic acid butyl ester,butyl chlorocarbonate,n-butyl chlorocarbonate,formic acid, chloro-, butyl ester,butyl-chloroformate,butylchloro-formate
61140
butyl carbonochloridate
CCCCOC(=O)Cl
Coupling reagent
Viscosity 0.95 mPa.s (20°C)
Packaging Glass bottle
Infrared Spectrum Authentic
Boiling Point 142.0°C
Color Undesignated
Density 1.0600g/mL
Melting Point -70.0°C
Quantity 1L
Refractive Index 1.4110 to 1.4130
Specific Gravity 1.06
Flash Point 39°C
Assay Percent Range 97.5% min. (GC)
Linear Formula ClCO2(CH2)3CH3
Beilstein 03, II, 11
Solubility Information Solubility in water: decomposes in water. Other solubilities: soluble in butanol,ether,benzol,soluble in most common organic solvents
Formula Weight 136.58
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Butyl chloroformate, 98%
Show More Show Less

GHS H Statement:
Toxic if inhaled.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-750-5

TSCA : TSCA