Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl methacrylate, 99%, stabilized, ACROS Organics™

Click to view available options
Quantity:
1L
5L
Packaging:
Glass bottle
Plastic drum
97-88-1,150-76-5
C8H14O2
142.2
MFCD00009444
SOGAXMICEFXMKE-UHFFFAOYSA-N
butyl methacrylate,n-butyl methacrylate,butylmethacrylate,2-propenoic acid, 2-methyl-, butyl ester,butyl 2-methacrylate,butylmethacrylaat,2-methyl-butylacrylat,butil metacrilato,2-methyl-butylacrylaat,2-methyl-butylacrylate
7354
butyl 2-methylprop-2-enoate
CCCCOC(=O)C(=C)C
97-88-1,150-76-5
C8H14O2
142.2
MFCD00009444
SOGAXMICEFXMKE-UHFFFAOYSA-N
butyl methacrylate,n-butyl methacrylate,butylmethacrylate,2-propenoic acid, 2-methyl-, butyl ester,butyl 2-methacrylate,butylmethacrylaat,2-methyl-butylacrylat,butil metacrilato,2-methyl-butylacrylaat,2-methyl-butylacrylate
7354
butyl 2-methylprop-2-enoate
CCCCOC(=O)C(=C)C
Viscosity 1.1 mPa.s (20°C)
Refractive Index 1.4225 to 1.4245
Specific Gravity 0.89
Stabilizer 10 to 110ppm MEHQ
Melting Point -75.0°C
Density 0.8900g/mL
Boiling Point 160.0°C to 163.0°C
Packaging Plastic drum
Infrared Spectrum Authentic
Quantity 5L
Color Undesignated
Flash Point 41°C
Assay Percent Range 98.5% min. (GC)
Linear Formula H2C=C(CH3)CO2(CH2)3CH3
Beilstein 02, III, 1286
Solubility Information Solubility in water: 3g/L in water (20°C). Other solubilities: very soluble in ether and ethanol,miscible in ethyl acetate
Formula Weight 142.2
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Butyl methacrylate, Stabilized
Show More Show Less

GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-615-1