Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl p-toluenesulfonate, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
778-28-9
C11H16O3S
228.306
MFCD00027203
QYJXDIUNDMRLAO-UHFFFAOYSA-N
butyl p-toluenesulfonate,butyl tosylate,n-butyl p-toluenesulfonate,butyl p-methylbenzenesulfonate,butyl-p-toluenesulfonate,p-toluenesulfonic acid, butyl ester,benzenesulfonic acid, 4-methyl-, butyl ester,butyl toluene-4-sulphonate,p-toluenesulfonic acid n-butyl ester,p-toluene sulfonic acid, butyl ester
13066
butyl 4-methylbenzenesulfonate
CCCCOS(=O)(=O)C1=CC=C(C=C1)C
778-28-9
C11H16O3S
228.306
MFCD00027203
QYJXDIUNDMRLAO-UHFFFAOYSA-N
butyl p-toluenesulfonate,butyl tosylate,n-butyl p-toluenesulfonate,butyl p-methylbenzenesulfonate,butyl-p-toluenesulfonate,p-toluenesulfonic acid, butyl ester,benzenesulfonic acid, 4-methyl-, butyl ester,butyl toluene-4-sulphonate,p-toluenesulfonic acid n-butyl ester,p-toluene sulfonic acid, butyl ester
13066
butyl 4-methylbenzenesulfonate
CCCCOS(=O)(=O)C1=CC=C(C=C1)C
Quantity 500g
Refractive Index 1.5030 to 1.5050 (20°C, 589nm)
Infrared Spectrum Authentic
Color Brown to Yellow
Density 1.12g/mL
Packaging Glass bottle
Assay Percent Range 99%
Linear Formula CH3C6H4SO3(CH2)3CH3
Formula Weight 228.31
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Butyl p-toluenesulfonate
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.