Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Butyl Palmitate 95.0+%, TCI America™
Click to view available options
Quantity:
25 mL
500 mL
Specifications
Color | Yellow |
Boiling Point | 170°C |
Quantity | 500 mL |
Formula Weight | 312.54 |
Percent Purity | ≥95.0% (GC) |
Physical Form | Liquid |
Chemical Name or Material | Butyl Palmitate |
Safety and Handling
EINECSNumber : (2)-0798
TSCA : Yes