Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Butyl-S Sepharose™ 6 Fast Flow Chromatography Media

The least hydrophobic medium in the Sepharose Fast Flow hydrophobic interaction chromatography (HIC) range and is particularly useful during the initial stages of purification process

$978.00

Specifications

Particle Size 45 - 165 µm
Product Type Butyl-S Sepharose 6 fast flow media
Flow Rate 250 to 400 cm/Hoµr
For Use With (Application) For Hydrophobic Medium in The Sepharose Fast Flow Hydrophobic Interaction Chromatography (HIC) Range and is Particularly Useful During The Initial Stages of PurifiCatIon Process
Ligand Type Butyl-S
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
45-002-941
View Documents Promotion Details
Cytiva
17097802
200 mL
Each for $978.00
Add to cart
 
Description

Description

  • Binds and elutes relatively strong hydrophobic molecules at comparatively low salt concentrations
  • Hydrophilic nature of the base matrix ensures low levels of non-specific binding
  • Purifies recombinant Hepatitis B virus surface antigen from CHO cells
  • Composed of cross-linked 6% agarose beads modified with butyl groups via thio-ether linkage
  • Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development

Specifications

Specifications

45 - 165 µm
250 to 400 cm/Hoµr
Butyl-S
100 kPa
Stable in commonly used aqueous Buffers: 1 mM HCl, 1 M NaOH, 30% Isopropanol, 50% Ethylene Glycol, 70% Ethanol, 6 M Guanidine hydrochloride, 8 M Urea
4°C to 30°C
Butyl-S Sepharose 6 fast flow media
For Hydrophobic Medium in The Sepharose Fast Flow Hydrophobic Interaction Chromatography (HIC) Range and is Particularly Useful During The Initial Stages of PurifiCatIon Process
6% Cross-Linked Agarose
3 to 13
20% Ethanol
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit