Learn More
Butylated Hydroxyanisole, NF, 98.5%, Spectrum™
Click to view available options
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
Quantity:
125 g
2.5 kg
B1194, 25013-16-5, C11H16O2
Spectrum™ Butylated Hydroxyanisole, NF is used for its antioxidant properties in pharmaceutical gels, creams, liquids, gelatin capsules and tablets. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle