Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butylated Hydroxyanisole, NF, 98.5%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
25013-16-5
C11H16O2
180.25
MRBKEAMVRSLQPH-UHFFFAOYSA-N
2-tert-butyl-4-methoxyphenol
COC1=CC=C(O)C(=C1)C(C)(C)C
25013-16-5
C11H16O2
180.25
MRBKEAMVRSLQPH-UHFFFAOYSA-N
2-tert-butyl-4-methoxyphenol
COC1=CC=C(O)C(=C1)C(C)(C)C

B1194, 25013-16-5, C11H16O2

Spectrum™ Butylated Hydroxyanisole, NF is used for its antioxidant properties in pharmaceutical gels, creams, liquids, gelatin capsules and tablets. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Quantity 45 kg
Ignition Residue 0.01%
Packaging Fiber Drum
CAS Max % 100%
Percent Purity 98.5%