Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Butylated Hydroxyanisole, NF, 98.5%, Spectrum™
Click to view available options
Quantity:
125 g
500 g
2.5 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
Description
Specifications
Ignition Residue | 0.01% |
Packaging | Amber Glass Bottle |
Quantity | 125 g |
CAS Max % | 100% |
Percent Purity | 98.5% |
Grade | NF |