Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butylated Hydroxytoluene, Granular, NF, 99-101.5%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
128-37-0
C15H24O
220.36
NLZUEZXRPGMBCV-UHFFFAOYSA-N
2,6-di-tert-butyl-4-methylphenol
CC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C
128-37-0
C15H24O
220.36
NLZUEZXRPGMBCV-UHFFFAOYSA-N
2,6-di-tert-butyl-4-methylphenol
CC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C

B1196, 128-37-0, C15H24O

Spectrum™ Butylated Hydroxytoluene, Granular, NF is used for its antioxidant properties in pharmaceutical gels, creams, liquids, gelatin capsules and tablets. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Ignition Residue 2e-005
Quantity 125 g
CAS Max % 1
Percent Purity 99 to 101.5%