Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butylbenzene, ≥99.8% (GC), MilliporeSigma™ Supelco™

Butylbenzene is a colourless organic solvent. It is used in pesticide manufacturing, plasticizer and also as solvent for coating. It is sometimes used as surface active agent, polymer linking agent and in organic synthesis.

$59.80 - $107.80

Specifications

CAS 104-51-8
Percent Purity ≥99.8% (GC)
Density 0.86 g/mL (at 25°C)
Grade Analytical Standard
Linear Formula C6H5(CH2)3 CH3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-9909
View Documents
MilliporeSigma Supelco
196005ML
5 mL
Each for $59.80
Add to cart
 
11-101-9075
View Documents
MilliporeSigma Supelco
1960010ML
10 mL
Each for $107.80
Add to cart
 
Description

Description

Featured Industries: Environmental
Specifications

Specifications

104-51-8
0.86 g/mL (at 25°C)
C6H5(CH2)3 CH3
≥99.8% (GC)
Analytical Standard
Neat
Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit