Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butylparaben, NF, 98-102%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
94-26-8
C11H14O3
194.23
QFOHBWFCKVYLES-UHFFFAOYSA-N
butyl 4-hydroxybenzoate
CCCCOC(=O)C1=CC=C(O)C=C1
94-26-8
C11H14O3
194.23
QFOHBWFCKVYLES-UHFFFAOYSA-N
butyl 4-hydroxybenzoate
CCCCOC(=O)C1=CC=C(O)C=C1

BU115, 94-26-8, C11H14O3

Spectrum™ Butylparaben, NF is an antimicrobial preservative used as a suspending agent for medications. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Ignition Residue 0.1%
Quantity 500 g
Packaging Poly Bottle
CAS Max % 100%
Percent Purity 98-102%