Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyltin trichloride, 97%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
1118-46-3
C4H9Cl3Sn
282.16
MFCD00000515
YMLFYGFCXGNERH-UHFFFAOYSA-K
butyltin trichloride,stannane, butyltrichloro,monobutyltin trichloride,butyltrichlorotin,trichlorobutyltin,butyl trichloro stannane,n-butyltin trichloride,trichlorobutylstannane,stannane, trichlorobutyl,monotributyltin trichloride
14236
butyl(trichloro)stannane
CCCC[Sn](Cl)(Cl)Cl
1118-46-3
C4H9Cl3Sn
282.16
MFCD00000515
YMLFYGFCXGNERH-UHFFFAOYSA-K
butyltin trichloride,stannane, butyltrichloro,monobutyltin trichloride,butyltrichlorotin,trichlorobutyltin,butyl trichloro stannane,n-butyltin trichloride,trichlorobutylstannane,stannane, trichlorobutyl,monotributyltin trichloride
14236
butyl(trichloro)stannane
CCCC[Sn](Cl)(Cl)Cl
Specific Gravity 1.69
Packaging Glass bottle
Flash Point 81°C
Density 1.6900g/mL
Infrared Spectrum Authentic
Boiling Point 93.0°C (10.0 mmHg)
Quantity 100g
Color Colorless to Yellow or Straw-Yellow
Refractive Index 1.5215 to 1.5245
Assay Percent Range 97%
Linear Formula CH3(CH2)3SnCl3
Beilstein 04, IV, 4346
Merck Index 15, 1594
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in most common organic solvents
Formula Weight 282.16
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Butyltin trichloride, 97%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Corrosive to the respiratory tract.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 214-263-6

RTECSNumber : WH6780000

TSCA : TSCA