Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyltin trichloride, 97%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
1118-46-3
C4H9Cl3Sn
282.16
MFCD00000515
YMLFYGFCXGNERH-UHFFFAOYSA-K
butyltin trichloride,stannane, butyltrichloro,monobutyltin trichloride,butyltrichlorotin,trichlorobutyltin,butyl trichloro stannane,n-butyltin trichloride,trichlorobutylstannane,stannane, trichlorobutyl,monotributyltin trichloride
14236
butyl(trichloro)stannane
CCCC[Sn](Cl)(Cl)Cl
1118-46-3
C4H9Cl3Sn
282.16
MFCD00000515
YMLFYGFCXGNERH-UHFFFAOYSA-K
butyltin trichloride,stannane, butyltrichloro,monobutyltin trichloride,butyltrichlorotin,trichlorobutyltin,butyl trichloro stannane,n-butyltin trichloride,trichlorobutylstannane,stannane, trichlorobutyl,monotributyltin trichloride
14236
butyl(trichloro)stannane
CCCC[Sn](Cl)(Cl)Cl
Specific Gravity 1.69
Flash Point 81°C
Refractive Index 1.5215 to 1.5245
Density 1.6900g/mL
Quantity 500g
Packaging Glass bottle
Color Colorless to Yellow or Straw-Yellow
Infrared Spectrum Authentic
Boiling Point 93.0°C (10.0 mmHg)
Assay Percent Range 97%
Linear Formula CH3(CH2)3SnCl3
Beilstein 04, IV, 4346
Merck Index 15, 1594
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in most common organic solvents
Formula Weight 282.16
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Butyltin trichloride, 97%
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 214-263-6

RTECSNumber : WH6780000

TSCA : TSCA