Alfa Aesar™ Butyraldoxime, 98%

Manufacturer: Alfa Aesar™ A1142036

Quantity 500g

 View more versions of this product

Catalog No. AAA1142036

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 110-69-0
    Molecular Formula C4H9NO
    Linear Formula CH3CH2CH2CH=NOH
    Formula Weight 87.12
    MDL Number MFCD00013943
    UN Number UN2840
    Beilstein 1698799
    Synonym Butanal oxime; Butyraldehyde oxime
    Chemical Name or Material Butyraldoxime

    Specifications

    Formula Weight 87.12
    Health Hazard 3 GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Synonym Butanal oxime; Butyraldehyde oxime
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H311-H226-H302-H319
    Toxic in contact with skin.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Causes serious eye irritation.
    Quantity 500g
    TSCA TSCA
    Linear Formula CH3CH2CH2CH=NOH
    MDL Number MFCD00013943
    CAS 110-69-0
    UN Number UN2840
    Beilstein 1698799
    Boiling Point 150° to 152°C
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTYRALDOXIME
    Molecular Formula C4H9NO
    Melting Point −29°C
    RTECS Number ES3500000
    Flash Point 58°C (136°F)
    Chemical Name or Material Butyraldoxime
    Assay 98%

    GHS H Statement
    H311-H226-H302-H319
    Toxic in contact with skin.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Causes serious eye irritation.

    GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit