Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyric acid hydrazide, 95%, Acros Organics™

Click to view available options
Quantity:
5g
25g
3538-65-6
C4H10N2O
102.137
FCCCRBDJBTVFSJ-UHFFFAOYSA-N
butyrohydrazide,butyric acid hydrazide,butanohydrazide,butyrylhydrazine,butanoic acid, hydrazide,n-butyric acid hydrazide,butyric hydrazide,butryic acid hydrazide,butanoic acid hydrazide,butyryl hydrazine
77079
butanehydrazide
CCCC(=O)NN
3538-65-6
C4H10N2O
102.137
FCCCRBDJBTVFSJ-UHFFFAOYSA-N
butyrohydrazide,butyric acid hydrazide,butanohydrazide,butyrylhydrazine,butanoic acid, hydrazide,n-butyric acid hydrazide,butyric hydrazide,butryic acid hydrazide,butanoic acid hydrazide,butyryl hydrazine
77079
butanehydrazide
CCCC(=O)NN
Infrared Spectrum Authentic
Quantity 5g
Boiling Point 120.0°C (10.0 mmHg)
Melting Point 43.0°C to 44.0°C
Linear Formula CH3CH2CH2C(O)NHNH2
Solubility Information Solubility in water: soluble.
Formula Weight 102.14
Percent Purity 95%
Chemical Name or Material Butyric acid hydrazide

GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning