Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyroin, 97%, Acros Organics™

Click to view available options
Quantity:
5g
25g
496-77-5
C8H16O2
144.22
BVEYJWQCMOVMAR-UHFFFAOYSA-N
butyroin,5-hydroxy-4-octanone,4-octanone, 5-hydroxy,5-octanol-4-one,octan-4-ol-5-one,fema no. 2587,5-hydroxy-octan-4-one,acmc-1aijr,ksc236m9n,+/--5-hydroxy-4-octanone
219794
5-hydroxyoctan-4-one
CCCC(C(=O)CCC)O
496-77-5
C8H16O2
144.22
BVEYJWQCMOVMAR-UHFFFAOYSA-N
butyroin,5-hydroxy-4-octanone,4-octanone, 5-hydroxy,5-octanol-4-one,octan-4-ol-5-one,fema no. 2587,5-hydroxy-octan-4-one,acmc-1aijr,ksc236m9n,+/--5-hydroxy-4-octanone
219794
5-hydroxyoctan-4-one
CCCC(C(=O)CCC)O
Boiling Point 80.0°C to 86.0°C (12.0mmHg)
Quantity 25g
Flash Point 71°C
Assay Percent Range 96% min. (GC)
Formula Weight 144.22
Percent Purity 97%
Chemical Name or Material Butyroin

GHS Signal Word: Warning

EINECSNumber : 207-830-4