Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyryl chloride 99%, ACROS Organics™

Click to view available options
Quantity:
50mL
250mL
1L
Packaging:
Glass bottle
141-75-3
C4H7ClO
106.55
MFCD00000752
DVECBJCOGJRVPX-UHFFFAOYSA-N
butyryl chloride,n-butyryl chloride,butyric acid chloride,butyroyl chloride,butyric chloride,butyrylchloride,unii-2xvm8e16ir,ccris 5989,n-butyric acid chloride,n-butanoyl chloride
8855
butanoyl chloride
CCCC(=O)Cl
141-75-3
C4H7ClO
106.55
MFCD00000752
DVECBJCOGJRVPX-UHFFFAOYSA-N
butyryl chloride,n-butyryl chloride,butyric acid chloride,butyroyl chloride,butyric chloride,butyrylchloride,unii-2xvm8e16ir,ccris 5989,n-butyric acid chloride,n-butanoyl chloride
8855
butanoyl chloride
CCCC(=O)Cl
Specific Gravity 1.018
Melting Point -89.0°C
Refractive Index 1.4112 to 1.4132
Density 1.0180g/mL
Quantity 50mL
Color Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Flash Point 18°C
Boiling Point 102.0°C
Assay Percent Range 99%
Linear Formula CH3CH2CH2COCl
Beilstein 02, 274
Merck Index 15, 1602
Solubility Information Solubility in water: decomposes. Other solubilities: miscible with ether,decomposes in alcohol
Formula Weight 106.55
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Butyryl chloride
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Toxic if inhaled.

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger