Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyrylcholinesterase/BCHE Mouse anti-Canine, Clone: OTI1E1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP2663810.1MG

Catalog No. NBP26638101


Add to cart

Description

Description

Butyrylcholinesterase/BCHE Monoclonal antibody specifically detects Butyrylcholinesterase/BCHE in Canine samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

Butyrylcholinesterase/BCHE
OTI1E1
Western Blot 1:500 - 1:2000, Flow Cytometry 1:100
Affinity Purified
BCHE
Full length human recombinant protein of human BCHE (NP_000046) produced in HEK293T cell.
Immunogen affinity purified
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Canine, Human
Flow Cytometry, Western Blot
Unconjugated
PBS pH 7.3 with 0.02% Sodium Azide
Acylcholine acylhydrolase, Butyrylcholine esterase, butyrylcholinesterase, CHE1cholinesterase, Choline esterase II, cholinesterase 1, E1, EC 3.1.1.8, Pseudocholinesterase
Mouse
IgG1
0.1mg
Primary
590
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit