Alfa Aesar™ Butyrylferrocene, 99%

Manufacturer: Alfa Aesar™ 3940418

Quantity 50g

 View more versions of this product

Catalog No. AA3940418

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1271-94-9
    Formula Weight 256.13
    MDL Number MFCD00143883
    Synonym Butyroferrocene; Ferrocenyl n-propyl ketone
    Chemical Name or Material Butyrylferrocene

    Specifications

    Formula Weight 256.13
    Physical Form Solid
    MDL Number MFCD00143883
    Boiling Point 329°C
    Synonym Butyroferrocene; Ferrocenyl n-propyl ketone
    Density 1.254g/mL
    Melting Point 36° to 40°C
    Quantity 50g
    CAS 1271-94-9
    Chemical Name or Material Butyrylferrocene
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit