Learn More
BiolegendSupplier Diversity Partner Brilliant Violet 510™ anti-human CD15 (SSEA-1)  DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 323028
Brilliant Violet 510™ anti-human CD15 (SSEA-1) [W6D3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-712-032