Learn More
BiolegendSupplier Diversity Partner Brilliant Violet 510™ anti-human CD183 (CXCR3)  DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 353726
Brilliant Violet 510™ anti-human CD183 (CXCR3) [G025H7]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-712-138