Learn More
BiolegendSupplier Diversity Partner Brilliant Violet 605™ anti-human CD8a  DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 301040
Brilliant Violet 605™ anti-human CD8a [RPA-T8]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus, Pigtailed Macaque, Sooty Mangabey; Apps: FC; Size: 100 tests
Catalog No. 50-711-961