Learn More
BiolegendSupplier Diversity Partner Brilliant Violet 711™ anti-human CD11b  DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 301343
Brilliant Violet 711™ anti-human CD11b [ICRF44]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Common Marmoset, Cynomolgus, Rhesus, Swine (Pig, Porcine); Apps: FC; Size: 25 tests
Catalog No. 50-711-971