Learn More
BiolegendSupplier Diversity Partner Brilliant Violet 711™ anti-human CD45  DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 304049
Brilliant Violet 711™ anti-human CD45 [HI30]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 25 tests
Catalog No. 50-711-986