Learn More
BiolegendSupplier Diversity Partner Brilliant Violet 785™ anti-human CD127 (IL-7Rα)  DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 351330
Brilliant Violet 785™ anti-human CD127 (IL-7Rα) [A019D5]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-712-124