Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BX 471, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent, selective non-peptide CCR1 antagonist (Ki = 1 nM for human CCR1). Exhibits 250-fold selectivity for CCR1 over CCR2, CCR5 and CXCR4. Inhibits MIP-α/CCL3-induced intracellular Ca2+ mobilization. Orally active; effectively reduces disease severity in a rat model of multiple sclerosis. Decreases renal fibrosis in a mouse model of obstructive nephropathy.
Specifications
Quantity | 10 mg |
Formula Weight | 434.89 |
Percent Purity | >98% |
Chemical Name or Material | BX 471 |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction