Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BYK 191023 dihydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and selective inhibitor of inducible nitric oxide synthase (iNOS) (IC50 values are 86, 17000, 162000 nM for iNOS, nNOS and eNOS respectively). Acts in an NADPH- and time-dependent manner. Active in vivo, reverses pathological hypotension in the rodent endotoxin model.
Specifications
Quantity | 50 mg |
Formula Weight | 327.21 |
Percent Purity | >99% |
Chemical Name or Material | BYK 191023 dihydrochloride |
Safety and Handling
Recommended Storage : Store at +4°C