Learn More
Tocris Bioscience™ C 21
Selective PRMT1 inhibitor
Supplier:  Tocris Bioscience™ 5128/1
Catalog No. 51-281
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Description
Description
C 21 is a selective protein arginine methyltransferase 1 (PRMT1) inhibitor (IC50 = 1.8 μM). Exhibits five-fold selectivity for PRMT1 over PRMT6 and >250-fold selectivity over PRMT3 and CARM1.
Specifications
Specifications
1 mg
C 21
2166.94
C 21
Product Suggestions
Product Suggestions