Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ c-Abl Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, and immunoprecipitation procedures.

Specifications

Antigen c-Abl Ab-1
Clone 8E9
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 6
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS938B
View Documents
Thermo Scientific™
MS938B
Biotinylated Ab with BSA and Azide 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS938P0
View Documents
Thermo Scientific™
MS938P0
Purified Ab with BSA and Azide 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS938P1
View Documents
Thermo Scientific™
MS938P1
Purified Ab with BSA and Azide 500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS938R1
View Documents
Thermo Scientific™
MS938R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS938B0
View Documents
Thermo Scientific™
MS938B0
Biotinylated Ab with BSA and Azide 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS938B1
View Documents
Thermo Scientific™
MS938B1
Biotinylated Ab with BSA and Azide 500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, and immunoprecipitation procedures with Thermo Scientific™ c-Abl Ab-1, Mouse Monoclonal Antibody.

The c-Abl proto-oncogene encodes a protein tyrosine kinase that is located in the cytoplasm and nucleus. In chronic myelogenous leukemia and in a subset of acute lymphoblastic leukemias, the c-Abl proto-oncogene undergoes a (9;22) chromosomal translocation producing a novel rearranged chromosome (the Philadelphia chromosome). As the result of the fusion of c-Abl sequences from chromosome 9 to the Bcr gene on chromosome 22. The molecular consequence of this translocation is the generation of a chimeric Bcr/Abl mRNA encoding activated Abl protein tyrosine kinase.

Host Species: Mouse

Clone: 8E9

Isotype: IgG1/γ

Species Reactivity: Human and Mouse. Others not tested.

Epitope: SH2 domain

Immunogen: Recombinant Abl protein

Molecular Weight: 120kDa

Positive Control: HeLa cells or placenta

Cellular Localization: Cytoplasmic and nuclear

Ab-1 reacts with c-Abl p120 and chimeric Bcr/Abl proteins.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

c-Abl Ab-1
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Murine
Signal Transduction
Recombinant Abl protein
8E9
Monoclonal
RUO
Primary
IgG1
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit