Learn More
c-Fos Monoclonal Antibody (4A3), PerCP, Invitrogen™
Mouse Monoclonal Antibody
Supplier:  Thermo Scientific MA545885
Encompass
Catalog No. PIMA545885
Add to Cart
This item is not returnable. View return policy
Description
Description
c-Fos Monoclonal antibody specifically detects c-Fos in Human, Rat samples. It is validated for Immunohistochemistry, Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
c-Fos
Monoclonal
1 mg/mL
2.15mM MES, 47.35mM phosphate with 1.033mM EDTA and no preservative; pH 7.4
P01100, P12841
FOS
Synthetic peptide from the N-terminal of Human c-Fos (aa. 280-380)
100 μg
Primary
Human, Rat
Liquid
Immunohistochemistry, Western Blot
4A3
PerCP
FOS
activator protein 1; AP-1; cellular oncogene c-fos; cellular oncogene fos; cFos; C-FOS; c-fos oncogene; c-fos protooncogene protein; D12Rfj1; FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS); FBJ murine osteosarcoma viral oncogene homolog; FBJ murine osteosarcoma viral oncogene homolog B; FBJ osteosarcoma oncogene; FBJ osteosarcoma oncogene B; FBJ osteosarcoma viral oncogene isoform deltaFosb-2; FOS; Fos proto-oncogene, AP-1 trancription factor subunit; Fos proto-oncogene, AP-1 transcription factor subunit; Fosb; FosB proto-oncogene, AP-1 transcription factor subunit; fra-2; G0/G1 switch regulatory protein 7; G0S7; I79_009428; p55; protein fosB; proto-oncogene cFOS; proto-oncogene c-Fos; proto-oncogene protein c-fos; v-fos FBJ murine osteosarcoma viral oncogene homolog
Mouse
Protein G
RUO
2353, 314322
4° C
IgG2
Recommendations
Recommendations