Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Fos Mouse anti-Human, Janelia Fluor™ 646, Clone: OTI7D6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270764JF646

Catalog No. N270764J646


Add to cart

Description

Description

c-Fos Monoclonal antibody specifically detects c-Fos in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

c-Fos
OTI7D6
Immunohistochemistry
Affinity Purified, Conjugated
FOS
Full length recombinant protein of human c-Fos (NP_005243) produced in E.coli.
Immunogen affinity purified
Apoptosis, Cancer, Cellular Markers, Hypoxia, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors
Primary
2353.0
Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
activator protein 1, AP-1, cellular oncogene c-fos, Cellular oncogene fos, c-fos, FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS), FBJ murine osteosarcoma viral oncogene homolog, G0/G1 switch regulatory protein 7, G0S7, proto-oncogene c-Fos, v-fos FBJ murine osteosarcoma viral oncogene homolog
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit