Learn More
c-Fos Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA124330
Description
PA1-24330 detects Fos in Human samples. PA1-24330 has been successfully used in Western Blot procedures. PA1-24330 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human FOS. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
The Fos gene family consists of 4 members: FOS, FOSB, FOSL1, and FOSL2. These genes encode leucine zipper proteins that can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. As such, the FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation. In some cases, expression of the FOS gene has also been associated with apoptotic cell death.
Specifications
| c-Fos | |
| Polyclonal | |
| Unconjugated | |
| P01100 | |
| FOS | |
| Synthetic peptide region of human FOS. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| FOS | |
| AP-1, C-FOS | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| 2353 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.