Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-jun Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI3G2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271059AF405

Catalog No. N271059A405


Add to cart

Description

Description

c-jun Monoclonal antibody specifically detects c-jun in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

c-jun
OTI3G2
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
JUN
Full length recombinant protein of human Jun (NP_002219) produced in HEK293T cells.
Immunogen affinity purified
Apoptosis, Cancer, Cell Cycle and Replication, Hypoxia, Immunology, Innate Immunity, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors, Wnt Signaling Pathway
Primary
3725.0
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
Activator protein 1, AP1, AP-1, c-Jun, enhancer-binding protein AP1, Jun activation domain binding protein, jun oncogene, jun proto-oncogene, Proto-oncogene c-Jun, transcription factor AP-1, V-jun avian sarcoma virus 17 oncogene homologp39, v-jun sarcoma virus 17 oncogene homolog, v-jun sarcoma virus 17 oncogene homolog (avian)
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit