Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
c-jun Mouse anti-Human, Mouse, Rat, APC, Clone: OTI3G2, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271059APC
Description
c-jun Monoclonal specifically detects c-jun in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
c-jun | |
Monoclonal | |
APC | |
PBS with 0.05% Sodium Azide | |
JUN | |
Full length recombinant protein of human Jun (NP_002219) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Flow Cytometry | |
OTI3G2 | |
Flow Cytometry | |
Activator protein 1, AP1, AP-1, c-Jun, enhancer-binding protein AP1, Jun activation domain binding protein, jun oncogene, jun proto-oncogene, Proto-oncogene c-Jun, transcription factor AP-1, V-jun avian sarcoma virus 17 oncogene homologp39, v-jun sarcoma virus 17 oncogene homolog, v-jun sarcoma virus 17 oncogene homolog (avian) | |
Mouse | |
Affinity Purified | |
Apoptosis, Cancer, Cell Cycle and Replication, Hypoxia, Immunology, Innate Immunity, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors, Wnt Signaling Pathway | |
3725.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title