Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C-Jun Mouse anti-Human, Clone: OTI3G2, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA500842

Catalog No. 50-167-1988


Add to cart

Description

Description

c-Jun is a transcription factor that recognizes and binds to the enhancer heptamer motif 5 ft.-TGA[CG]TCA-3 ft.. It promotes activity of NR5A1 when phosphorylated by HIPK3 leading to increased steroidogenic gene expression upon cAMP signaling pathway stimulation. Involved in activated KRAS-mediated transcriptional activation of USP28 in colorectal cancer (CRC) cells.
Specifications

Specifications

C-Jun
Monoclonal
0.9 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P05412
AP-1, AP1, c-Jun, p39
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3725
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI3G2
Unconjugated
JUN
Liquid
JUN
Full length human recombit protein of human JUN produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit